MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit
Catalog # AATB22413
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:MycoLight™ rapid fluorescence bacterial gram stain kit
Specifications
About this item
The MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit provides an easy and convenient way for determination of gram sign in live bacteria.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimised for fluorescent microscopy
Caution: For research use only.