Order Entry
Austria
ContactUsLinkComponent
MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit
MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit
Catalog # AATB22405
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit
Catalog # AATB22405
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    MycoLight™ fluorimetric CTC live bacteria quantification kit

Specifications

About this item

The CTC Flow Cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial health and vitality as a function of the respiratory activity.

  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimised for flow cytometry
Caution: For research use only.