PSA™ iFluor® 488 Imaging Kit with Goat Anti-Mouse IgG
Catalog # AATB45260
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
Specifications
About this item
iFluor® 488 PSA kit is a much superior replacement for Alexa Fluor 488 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
- Higher efficiency
- Improved fluorescence signal
- Less consumption of primary antibody
- 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.