Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Mitochondrial Autophagy Imaging Kit *Red Fluorescence*
Cell Meter™ Mitochondrial Autophagy Imaging Kit *Red Fluorescence*
Catalog # AATB22998
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Mitochondrial Autophagy Imaging Kit *Red Fluorescence*
Catalog # AATB22998
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ mitochondrial autophagy imaging kit *red fluorescence*
  • Tests per kit:
    100 Tests

Specifications

About this item

Mitochondrial autophagy (also called 'mitophagy') appears to be involved in Alzheimer and Parkinson diseases, mainly induced by the accumulation of depolarised mitochondria.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.