ReadiLink™ xtra Rapid Cy7 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Catalog # AATB1973
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ xtra rapid Cy7 antibody labelling kit 'BSA-compatible'
Specifications
About this item
ReadiLink™ xtra rapid antibody labelling kits require essentially only two simple mixing steps without a column purification needed.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labeling protocol
- No column purification required
Caution: For research use only.