Bucculite™ FdU Cu-Free Cell Proliferation Fluorescence Imaging Kits
Supplier: AAT Bioquest
Certificates
About this item
Monitoring cell proliferation is one of the most reliable methods to assess cell viability, cell cycles and genotoxicity.
- Environmentally friendly assay
- Robust protocol
- Stable signal
- Optimised for common channels
Related Information
Manual Bucculite Fdu Cu Free Cell Proliferation Fluorescence Imaging Kit Deep Red Fluorescence.pdf Manual Bucculite™ Fdu Cu Free Cell Proliferation Fluorescence Imaging Kit Green Fluorescence.pdf Manual Bucculite™ FdU Cu-Free Cell Proliferation Fluorescence Imaging Kit Red Fluorescence.pdf Manuals Bucculite™ FdU Cu-Free Cell Proliferation Fluorescence Imaging Kit.pdf
Manual
Frequently Bought Together