Cell Meter™ Fluorimetric Live Cell Cycle Assay Kit *Green Fluorescence Optimised for Flow Cytometry*
Catalog # AATB22841
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ fluorimetric live cell cycle assay kit *green fluorescence optimized for flow Cytometry*
Specifications
About this item
The Cell Meter™ assay kits are a set of tools for monitoring cell viability and proliferation.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.