ContactUsLinkComponent
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Red Fluorescence* (50 Tests)
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Red Fluorescence* (50 Tests)
  AATB22844
 :  
undefined
Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Red Fluorescence* (50 Tests)
  AATB22844
 :  
  • Pk:
    50 Tests
  • Description:
    Cell Meter™ live cell TUNEL apoptosis assay kit *red fluorescence*
DNA fragmentation represents a characteristic of late stage apoptosis.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
 : For research use only.