Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimised for Microplate Readers*
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimised for Microplate Readers*
Catalog # AATB22792
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimised for Microplate Readers*
Catalog # AATB22792
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ phosphatidylserine apoptosis assay kit *red fluorescence optimized for microplate readers*

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.