Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Solids Certified Reference Material
Solids Certified Reference Material
Solids Certified Reference Material
Catalog # ACCUWC-SOL
Supplier:
AccuStandard
CAS Number:
Expand image
Solids Certified Reference Material
Catalog # ACCUWC-SOL
Supplier:
AccuStandard
CAS Number:
Specifications
Pk:
100 mL
Description:
Solids certified reference material
Storage temperature:
>5 °C
Appearance:
Solid
Specifications
About this item
Product Details
Product Details