All-Weather Vinyl Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
: Brady
Versatile and durable label printer tape is compatible with BMP41/BMP51/BMP53/M511 printers.
- Versatile labels are ideal for labelling pipes, equipment, shelves, and other industrial surfaces
- Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
- Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
- Weather-resistant labels designed to last up to 10 years in an outdoor environment
- For material specific performance and properties, please refer to the Technical Data Sheet (TDS)
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label printer.
Gloss finish, abrasion-resistant, chemical-resistant, oil-resistant, water-resistant, outdoor use B-595 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
: BS 5609:1986 Section 2 and Section 3. RoHS compliant to 2011/65/EU (Pass 2015/863).
Frequently Bought Together