ContactUsLinkComponent
RNA model
  763-0455
undefined
RNA model
  763-0455
  • Description:
    RNA model
  • Pk:
    1
RNA 12 base single stranded model kit.
  • Two TRNA and two amino acid parts
  • Original, unique and dual-scale system
  • Can be mounted on the DNA-model
These sets are suitable for students but may also be used by scientists.