Order Entry
Austria
ContactUsLinkComponent
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml)
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml)
Catalog # AATB20011
Supplier:  AAT Bioquest
CAS Number:  
undefined
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml)
Catalog # AATB20011
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Hank's Balanced Salt Solution with 20 mm Hepes buffer (HHBS)

Specifications

About this item

Hank's Balanced Salt Solution with 20 mM Hepes buffer (HHBS) is used for a variety of cell culture applications, such as washing cells, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

  • Excellent buffering capacity
  • Highly cited
  • Protocol available