ContactUsLinkComponent
Transfectamine™ 5000 Transfection Reagent
Transfectamine™ 5000 Transfection Reagent
Supplier:  AAT BIOQUEST
Certificates
undefined
Transfectamine™ 5000 Transfection Reagent
Supplier:  AAT BIOQUEST
Specifications
  • Bioproduction:
    Yes
  • Cell specific:
    Mammalian
  • DNA and siRNA:
    Yes
  • DNA:
    Yes
  • shRNA:
    Yes
  • Storage temperature:
    Freeze (< –15 °C); minimise light exposure