DIG-HRP Conjugate (1 mg)
AATB11023
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:Digoxigenin
- Source:Eukaryotic cells
Digoxigenin (DIG)-conjugated horse radish peroxidase (HRP) is an excellent tool for biological detections with ELISA and other DIG-coupled assays.
- Selective binding
- ELISA development applications
: For research use only.