cBSA *Optimised for Maximum Immunisation Response* (10 mg)
AATB5600
- Serum format:Lyophilised
- Sterility:Non sterile
- Description:BSA cationized with ethylenediamine
- Storage temperature:<–20 °C
- Application:Use as a carrier protein for immunisation
- Purification:97%, via fractionation
- Pack type:Bottle
- Pk:10 mg
cBSA is the bovine serum albumin modified with ethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
- Lyophilized powder
- Excellent quality control
- Stronger immune response
- 60000 molecular weight
: For research use only.