ContactUsLinkComponent
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit)
  AATB20001
 :  
undefined
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit)
  AATB20001
 :  
  • Pk:
    1 KIT
  • Description:
    FluoroQuest™ anti-fading kit I *optimized for slide imaging*
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
 : For research use only.