Native Bovine Bovine Serum Albumin (from Serum), Semicarbazide (SEM)
Catalog # AATB50007
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Semicarbazide (SEM)
- Protein/peptide type:Native
- Source:Serum
- Species:Bovine
- Protein synonyms:BSA
- Protein/peptide name:Bovine Serum Albumin
Specifications
About this item
Semicarbazide (SEM)-BSA Conjugate is used for developing anti-Semicarbazide (SEM) antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Caution: For research use only.