Native Armoracia rusticana HRP (from Armoracia rusticana), 3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)
AATB50015
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)
- Protein/peptide type:Native
- Source:Armoracia rusticana
- Species:Armoracia rusticana
- Protein synonyms:Horseradish Peroxidase
- Protein/peptide name:HRP
3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)-HRP conjugate is used for developing an ELISA assay for quantifying 3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ).
- Assay development
- High sensitivity
: For research use only.