Order Entry
Austria
ContactUsLinkComponent
 
Native Armoracia rusticana HRP (from Armoracia rusticana), Chloramphenicol
  AATB50003
 :  
undefined
Native Armoracia rusticana HRP (from Armoracia rusticana), Chloramphenicol
  AATB50003
 :  

 

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Chloramphenicol
  • Protein/peptide type:
    Native
  • Source:
    Armoracia rusticana
  • Species:
    Armoracia rusticana
  • Protein synonyms:
    Horseradish Peroxidase
  • Protein/peptide name:
    HRP

 

 

Chloramphenicol-HRP conjugate is used for developing an ELISA assay for quantifying chloramphenicol.

  • Assay development
  • High sensitivity
 : For research use only.