Native Dextran (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Catalog # AATB21700
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Protein synonyms:Manninotriose|Macrodex
- Protein/peptide name:Dextran
Specifications
About this item
FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
- Well-retained in cells
- 20000 mw
- Multiple applications
Caution: For research use only.