ContactUsLinkComponent
PSA™ iFluor® 488 Imaging Kit with Goat Anti-Rabbit IgG
PSA™ iFluor® 488 Imaging Kit with Goat Anti-Rabbit IgG
  AATB45205
 :  
undefined
PSA™ iFluor® 488 Imaging Kit with Goat Anti-Rabbit IgG
  AATB45205
 :  
  • Pk:
    100 Tests
  • Description:
    Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
iFluor® 488 PSA kit is a much superior replacement for Alexa Fluor 488 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
 : Plastic vial.
 : For research use only.