Order Entry
Austria
ContactUsLinkComponent
ReadiLink™ Rapid iFluor® Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
ReadiLink™ Rapid iFluor® Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1250
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1250
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid iFluor™ 750 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'

Specifications

About this item

AAT Bioquest's iFluor® dyes are developed for labelling proteins, in particular, antibodies.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.