Order Entry
Austria
ContactUsLinkComponent
ReadiLink™ xtra Rapid XFD555 Antibody Labelling Kit *BSA-Compatible, XFD555 Same Structure to Alexa Fluor™ 555* (2 Labellings)
ReadiLink™ xtra Rapid XFD555 Antibody Labelling Kit *BSA-Compatible, XFD555 Same Structure to Alexa Fluor™ 555* (2 Labellings)
Catalog # AATB1980
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ xtra Rapid XFD555 Antibody Labelling Kit *BSA-Compatible, XFD555 Same Structure to Alexa Fluor™ 555* (2 Labellings)
Catalog # AATB1980
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ xtra rapid XFD555 antibody labeling kit 'BSA-compatible, XFD555 same structure to Alexa Fluor™ 555'

Specifications

About this item

XFD555 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 555 (Alexa Fluor® is the trademark of ThermoFisher).

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.