Buccutite™ ALP (Alkaline Phosphatase) Antibody Conjugation Kits (2 Labelings)
Protein-protein conjugations are commonly performed with a bifunctional linker (such as SMCC) having different reactivity at each end for linking two different proteins.
- High conjugation yield
- Stable crosslinking
- Minimal hands-on time
- Robust protocol
Manual ALP (Alkaline Phosphatase) Antibody Conjugation Kits (2 Labelings).pdf Manual Buccutite ALP Alkaline Phosphatase Antibody Conjugation Kit Optimized for Labeling 100 μg Protein.pdf Manual Buccutite Alp Alkaline Phosphatase Antibody Conjµgation Kit Optimized for Labeling 100 µg Protein.pdf Manual Buccutite™ ALP (Alkaline Phosphatase) Antibody Conjugation Kit *Optimized for Labeling 25 ug Protein.pdf Manual Buccutite™ ALP (Alkaline Phosphatase) Antibody Conjugation Kits.PDF
Frequently Bought Together