Order Entry
Austria
ContactUsLinkComponent
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml)
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml)
Catalog # AATB24100
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml)
Catalog # AATB24100
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Description:
    ReadiUse™ bacterial cell lysis buffer

Specifications

About this item

Cell lysis refers to the breaking down of cells, and it is often used to analyze specific cellular compositions such as proteins, lipids, nucleic acids, reporter molecules, cell signal molecules and other small biomolecules.

  • ReadiUse™ reagents require minimal hands-on time