ContactUsLinkComponent
Tetracysteine REASH Acetate *2 mM DMSO Solution*
Tetracysteine REASH Acetate *2 mM DMSO Solution*
Catalog # AATB22336
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Tetracysteine REASH Acetate *2 mM DMSO Solution*
Catalog # AATB22336
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Tetracysteine REASH acetate *2 mM DMSO solution*
  • Environmentally Preferable:
  • Tests per kit:
    100 Tests
Specifications
About this item
REASH acetate is a cell-permeable version of REASH, which is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.