Order Entry
Austria
ContactUsLinkComponent
MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit
MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit
Catalog # AATB22411
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit
Catalog # AATB22411
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    MycoLight™ Fluorescence live/dead bacterial Imaging kit
  • Tests per kit:
    100 Tests

Specifications

About this item

AAT Bioquest's MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit provides two-colour fluorescence assay for visualizing live and dead bacteria through fluorescent microscope.

  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for fluorescent microscopy
Caution: For research use only.