ReadiUse™ NADPH Regenerating Kit
Catalog # AATB15265
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:1.000 Tests
- Description:ReadiUse™ NADPH regenerating kit
- Tests per kit:1000 Tests
Specifications
About this item
NADPH provides the reducing equivalents for biosynthetic reactions and for oxidation-reduction involved in protection against the toxicity of ROS (reactive oxygen species).
- Ready to use
- Robust protocol
- Contains all necessary components
Caution: For research use only.