Amplite® Fluorimetric Beta-Galactosidase Assay Kit *Green Fluorescence*
E. coli beta-galactosidase is a 464 Kd tetramer.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.
Manual Amplite Fluorimetric Beta Galactosidase Assay Kit Red Fluorescence.pdf Manual Amplite™ Fluorimetric Beta Galactosidase Assay Kit Green Fluorescence.pdf Manual Fluorimetric Beta-Galactosidase Assay Kit Green Fluorescence.pdf Manual Fluorimetric Beta-Galactosidase Assay Kit Green Fluorescences.pdf
Frequently Bought Together