ContactUsLinkComponent
Amplite® Colorimetric Maleimide Quantitation Kit
Catalog # AATB5525
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Maleimide Quantitation Kit
Catalog # AATB5525
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ colorimetric maleimide quantitation kit
  • Tests per kit:
    100 Tests
Specifications
About this item
Maleimides can be directly assayed spectrophotometrically at 302 nm.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.