Amplite® Colorimetric Aldehyde Quantitation Kit *Blue Color*
Catalog # AATB10053
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ colorimetric aldehyde quantitation kit *blue color*
- Tests per kit:200 Tests
Specifications
About this item
The formation, reactivity and toxicity of aldehydes originating from the peroxidation of lipids of cellular membranes have received great attention in recent years.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.