ContactUsLinkComponent
Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Catalog # AATB10070
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Catalog # AATB10070
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric malondialdehyde (MDA) quantitation kit
  • Tests per kit:
    200 Tests
Specifications
About this item
Malondialdehyde (MDA) is natural byproduct of lipid peroxidation and is widely used as a key indicator to determine the oxidative stress and free radical formation.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.