Cell Meter™ Fluorimetric Live Cell Cycle Assay Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22860
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ Fluorimetric live cell cycle assay kit *red fluorescence optimized for flow Cytometry*
- Tests per kit:100 Tests
Specifications
About this item
Cell cycles have four sequential phases: G0/G1, S, G2, and M.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.