Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Red Fluorescence*
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Red Fluorescence*
Catalog # AATB22853
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Red Fluorescence*
Catalog # AATB22853
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 Tests
  • Description:
    Cell Meter™ Fixed cell and tissue TUNEL apoptosis assay kit *red fluorescence*
  • Tests per kit:
    25 Tests

Specifications

About this item

Cell Meter™ TUNEL apoptosis assay kit provides a robust tool for conveniently detecting apoptosis caused by DNA fragmentation.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.