Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*
Catalog # AATB22803
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*
Catalog # AATB22803
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    500 Tests
  • Description:
    Cell Meter™ NIR mitochondrion membrane potential assay kit *optimized for microplate reader*
  • Tests per kit:
    500 Tests

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimized for microplate readers but suitable for all major fluorescence platforms
Caution: For research use only.