Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Flow Cytometry*
Catalog # AATB22970
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ Fluorimetric mitochondrial superoxide activity assay kit*Optimized for flow cytometry*
- Tests per kit:100 Tests
Specifications
About this item
Mitochondria are major producers of cellular superoxide.
- Ready to use
- Contains all necessary components
- Robust and adaptable
- Optimized for flow cytometry but suitable for all major fluorescence platforms
- Results with only 1 hour incubation
Caution: For research use only.