Cell Meter™ Fluorimetric Fixed Cell Cycle Assay Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22842
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ Fluorimetric fixed cell cycle assay kit *Red fluorescence optimized for flow cytometry*
- Tests per kit:100 Tests
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability and proliferation.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.