ContactUsLinkComponent
Cell Navigator® Cell Plasma Membrane Staining Kit *Red Fluorescence*
Cell Navigator® Cell Plasma Membrane Staining Kit *Red Fluorescence*
Catalog # AATB22681
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Navigator® Cell Plasma Membrane Staining Kit *Red Fluorescence*
Catalog # AATB22681
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Cell Navigator™ Cell plasma membrane staining kit *Red fluorescence*
  • Tests per kit:
    500 Tests
Specifications
About this item
The Cell Navigator® Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins.
  • Signal persists through fixation
  • Stable labeling regardless of cell type
  • Multiple applications
Caution: For research use only.