Order Entry
Austria
ContactUsLinkComponent
Cell Explorer™ Live Cell Labelling Kit *Red Fluorescence*
Cell Explorer™ Live Cell Labelling Kit *Red Fluorescence*
Catalog # AATB22609
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Explorer™ Live Cell Labelling Kit *Red Fluorescence*
Catalog # AATB22609
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Explorer™ Live cell labeling Kit *Red fluorescence*
  • Tests per kit:
    200 Tests

Specifications

About this item

Our Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Caution: For research use only.