Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Apoptotic and Necrotic Multiplexing Detection Kit I *Triple Fluorescence Colors*
Cell Meter™ Apoptotic and Necrotic Multiplexing Detection Kit I *Triple Fluorescence Colors*
Catalog # AATB22840
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Apoptotic and Necrotic Multiplexing Detection Kit I *Triple Fluorescence Colors*
Catalog # AATB22840
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ apoptotic and necrotic multiplexing detection kit I *triple fluorescence colors*
  • Tests per kit:
    100 Tests

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.