Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry with 405 nm excitation*
Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry with 405 nm excitation*
Catalog # AATB22809
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry with 405 nm excitation*
Catalog # AATB22809
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ intracellular GSH assay kit *optimized for flow cytometry with 405 nm excitation*
  • Tests per kit:
    100 Tests

Specifications

About this item

There are a variety of parameters that can be used for monitoring cell apoptosis.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.