Cell Explorer™ Live Cell Tracking Kit *Deep Red Fluorescence*
Catalog # AATB22624
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Explorer™ live cell tracking kit *deep red fluorescence*
- Tests per kit:200 Tests
Specifications
About this item
The Cell Explorer™ fluorescence imaging kits are a set of tools for labeling cells for fluorescence microscopic investigations of cellular functions.
- Long label retention
- Suitable for all major fluorescence platforms
- Contains all necessary components
- Multiple applications
Caution: For research use only.