Cell Meter™ Fluorimetric Intracellular Nitric Oxide (NO) Activity Assay Kit *Orange Fluorescence Optimized for Flow Cytometry*
Catalog # AATB16351
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ Fluorimetric intracellular Nitric Oxide (NO) activity assay kit *Orange fluorescence optimized for flow cytometry*
- Tests per kit:100 Tests
Specifications
About this item
Nitric oxide (NO) is an important biological regulator involved in numbers of physiological and pathological processes.
- High sensitivity
- Bright fluorescence
- Suitable for live cell imaging
Caution: For research use only.