Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Green Fluorescence*
Artikelnr. AATB11503
Lieferant: AAT BIOQUEST
CAS-Nummer:
Spezifikationen
- Pk:200 Tests
- Description:Cell Meter™ intracellular fluorimetric hydrogen peroxide assay kit *Green Fluorescence*
- Tests per kit:200 Tests
Spezifikationen
Über diesen Artikel
Hydrogen peroxide (H₂O₂) is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
- Mix-and-read format
- Adaptable to hts instrumentation
- Quantifies h2o2 in live cells
Caution: For research use only.