ContactUsLinkComponent
Annexin V-mFluor™ Violet 540 Conjugate
Annexin V-mFluor™ Violet 540 Conjugate
Catalog # AATB20082
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Annexin V-mFluor™ Violet 540 Conjugate
Catalog # AATB20082
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Annexin V-mFluor™ Violet 540 conjugate
  • Tests per kit:
    100 Tests
Specifications
About this item
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
  • Optimized for flow cytometry
Caution: For research use only.