Helix Fluor™ 575 Succinimidyl Ester
Catalog # AATB251
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:1 mg
- Description:Helix Fluor™ 575 Succinimidyl Ester
- Purity:90%
Specifications
About this item
FAM, TET, VIC, JOE, NED, TAMRA and ROX fluorescent dyes are frequently used for labeling oliogs.
- Multiple Bioconjugation Applications
- Excellent Quality
Caution: For research use only.