Order Entry
Austria
ContactUsLinkComponent
Proteinase K Solution
Proteinase K Solution
Catalog # VIAG502-PK
Supplier:  VIAGEN BIOTECH
CAS Number:  
Proteinase K Solution
Catalog # VIAG502-PK
Supplier:  VIAGEN BIOTECH
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Form:
    Liquid
  • Shipping conditions:
    Approved for shipment on wet or dry ice
  • Purity:
    DNase-free, RNase-free
  • Concentration:
    20 mg/ml

Specifications

About this item

Proteinase K from the fungus engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins. Proteinase K remains active.

  • Over a wide pH range optimal activity between 6,5 and 9,5
  • Under denaturing conditions e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents e.g., EDTA
  • At comparatively high temperatures optimum digestion temperature is 65 °C

Removal of endogenous nucleases during the preparation of DNA and RNA, preparation of tissue sections for in situ hybridisation. Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Caution: For research use only. Not for use in diagnostic procedures.