
Lithium bromide, beige
Catalog # SIAL746479-100G
Supplier: Merck
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:86.84 g/mol
Boiling Pt:1.265 °C
Melting Pt:547 °C
Density:3,460 g/cm3 at 20 °C
:GHS07
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077