Green-Pak™ / SpaceSaver™ Pipette Tip Racks, Rainin
613-4700
: RAININ
- Colour code:Red
- Description:Green-Pak®, empty racks/lids
- Seq:50
- For use with:GP-10G, GPS-10G refills
- Pk:10
Empty rack/lid for refill with Green-Pak™ and SpaceSaver™ 10 μL universal-fit ShaftGard™ tips.